Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-003

Supelco

R(-)-Selegiline solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(-)-Deprenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N
CAS Number:
Molecular Weight:
187.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@@H](N(CC#C)C)CC1=CC=CC=C1

InChI

1S/C13H17N/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13/h1,5-9,12H,10-11H2,2-3H3/t12-/m1/s1

InChI key

MEZLKOACVSPNER-GFCCVEGCSA-N

Gene Information

human ... MAOB(4129)

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. Selegiline is an antidepressant, which belongs to the phenethylamine class of drugs. It is sold under the trade names Anipryl®, Emsam®, and Zelapar® for treatment of Parkinson′s disease, depression, and senile dementia.

Legal Information

Anipryl is a registered trademark of Deprenyl Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Emsam is a registered trademark of Somerset Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zelapar is a registered trademark of Valeant Pharmaceuticals North America LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wakako Maruyama et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(1), 83-89 (2012-08-16)
Neuroprotection has been proposed in neurodegenerative disorders, such as Parkinson's and Alzheimer's diseases, to delay or halt disease progression or reverse neuronal deterioration. The inhibitors of type B monoamine oxidase (MAO), rasagiline and (-)deprenyl, prevent neuronal loss in cellular and
Chi-Un Pae et al.
The Journal of clinical psychiatry, 73(5), 661-668 (2012-06-16)
The objective of this analysis is to present the safety profile of selegiline transdermal system (STS) in clinical practice after US Food and Drug Administration approval by analyzing reported postmarketing adverse events (AEs). Deidentified data were obtained on AEs, regardless
Retention rate of selegiline in early Parkinson's disease: a retrospective survey.
T Keränen et al.
International journal of clinical practice, 66(10), 1014-1014 (2012-09-22)
Chong Li et al.
Biochemistry. Biokhimiia, 77(6), 671-678 (2012-07-24)
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), a glycolytic enzyme, plays an important role in glycolysis. It was reported that GAPDH undergoes S-nitrosylation, which facilitated its binding to Siah1 and resulted in nuclear translocation and cell apoptosis. The results of this study show that
Jardel Gomes Villarinho et al.
European journal of pharmacology, 690(1-3), 107-114 (2012-07-10)
In this study we assessed the involvement of monoamine oxidase B (MAO-B), a key enzyme implicated in monoamine metabolism, on postoperative (plantar incision) and neuropathic (partial sciatic nerve ligation) pain models in mice. Paw incision submitted mice showed a significant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service