Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R-019

Supelco

Retigabine-D4 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14D4FN3O2
Molecular Weight:
307.36
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

FC1=C([2H])C([2H])=C(CNC2=CC(N)=C(NC(OCC)=O)C=C2)C([2H])=C1[2H]

InChI

1S/C16H18FN3O2/c1-2-22-16(21)20-15-8-7-13(9-14(15)18)19-10-11-3-5-12(17)6-4-11/h3-9,19H,2,10,18H2,1H3,(H,20,21)/i3D,4D,5D,6D

InChI key

PCOBBVZJEWWZFR-LNFUJOGGSA-N

General description

An internal standard and Certified Spiking Solution® suitable for quantitation of retigabine levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, urine drug testing, medication monitoring, or pharmaceutical research. Retigabine, also known as ezogabine, is a new antiepileptic drug developed for treatment of partial seizures. The drug is marketed under the trade names Potiga® in the US and Trobalt in the EU.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Potiga is a registered trademark of Valeant Pharmaceuticals North America LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

42.8 °F - closed cup - Solvent

flash_point_c

6.0 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service