Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Q-006

Supelco

Quazepam solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11ClF4N2S
CAS Number:
Molecular Weight:
386.79
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C17H11ClF4N2S/c18-10-5-6-14-12(7-10)16(11-3-1-2-4-13(11)19)23-8-15(25)24(14)9-17(20,21)22/h1-7H,8-9H2

Inchi Key

IKMPWMZBZSAONZ-UHFFFAOYSA-N

General description

Quazepam, sold as Doral and Dormalin, is a long-acting benzodiazepine derivative drug for the treatment of insomnia including sleep induction and sleep maintenance. A Certified Snap-N-Spike® Solution suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service