Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O-021

Supelco

Omeprazole solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Omeprazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Antra, Losec

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O3S
CAS Number:
Molecular Weight:
345.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(OC)c3C

InChI

1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)

InChI key

SUBDBMMJDZJVOS-UHFFFAOYSA-N

Gene Information

General description

Omeprazole, sold under the trade name Prilosec®, is a proton pump inhibitor used in the treatment of peptic ulcers and GERD. This Snap-N-Spike® reference solution is applicable for use in clinical toxicology or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Prilosec is a registered trademark of Astrazeneca AB
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James G Sanders et al.
The New Zealand medical journal, 125(1366), 68-73 (2012-12-21)
We present an atypical case of subglottic stenosis with diffuse tracheal stenoses in a child responsive only to steroid and azithromycin (AZI) therapy. A 12-year-old boy presented with acute biphasic stridor on the background of an 18-month history of progressive
E A Sorokina et al.
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology, (5)(5), 14-21 (2013-02-14)
The estimation of gastric contents pH and gastroduodenal zone mucous membrane condition based on the fibrogastroduodenoscopy data of40 patients who were examined and received proton pump inhibitors in the postoperative period after heart surgical intervention. It was estimated, that acidic
[Comment of the Delphi Care editors on Critical evaluation of interaction screening software available in pharmacies].
Daniel Duh et al.
Journal de pharmacie de Belgique, (4)(4), 14-15 (2013-01-29)
J Labenz et al.
MMW Fortschritte der Medizin, 154 Suppl 4, 110-114 (2013-01-19)
Heartburn and acid regurgitation--cardinal symptoms of gastro-esophageal reflux disease (GERD)--rank among the most frequent gastrointestinal disorders. The high degree of suffering often requires a fast and effective symptom relief. Proton pump inhibitors (PPI) are the medication of choice for all
V A Zolotarev et al.
Bulletin of experimental biology and medicine, 154(4), 415-418 (2013-03-15)
Proton pump inhibitors were shown to affect the sensitivity of the gastric mucosa to chemical agents. This effect is associated with inhibition of proton back-diffusion and increase in the permeability of the gastric epithelium. We studied the effect of omeprazole

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service