Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID990

6-(2-Aminopropyl)benzofuran hydrochloride

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO · HCl
CAS Number:
Molecular Weight:
211.69
UNSPSC Code:
41116107

agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C11H13NO.ClH/c1-8(12)6-9-2-3-10-4-5-13-11(10)7-9;/h2-5,7-8H,6,12H2,1H3;1H

InChI key

APEWOTOLPKNSPE-UHFFFAOYSA-N

General description

6-APB, also referred to by its street name Benzo Fury, is a stimulant with hallucinogenic effects similar to MDA. The designer drug and MDA analog is sometimes found in certain bath salt blends. This reference material is suitable as starting material in calibrators and controls for use in LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service