Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID940

Isometheptene mucate

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H48N2O8
CAS Number:
Molecular Weight:
492.65
UNSPSC Code:
41116107

grade

certified reference material

form

solid

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

InChI

1S/C9H19N.C6H10O8/c1-8(2)6-5-7-9(3)10-4;7-1(3(9)5(11)12)2(8)4(10)6(13)14/h6,9-10H,5,7H2,1-4H3;1-4,7-10H,(H,11,12)(H,13,14)

InChI key

HZEDCQLNSYIOTE-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service