Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID924

Methyl codeine

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO3
CAS Number:
Molecular Weight:
313.39
UNSPSC Code:
41116107

manufacturer/tradename

NMI Australia

format

neat

Storage temp.

room temp

InChI

1S/C19H23NO3/c1-20-9-8-19-12-5-7-15(22-3)18(19)23-17-14(21-2)6-4-11(16(17)19)10-13(12)20/h4-7,12-13,15,18H,8-10H2,1-3H3/t12-,13+,15-,18-,19-/m0/s1

Inchi Key

HGPQAWTZLJXCTC-SSTWWWIQSA-N

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service