Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID905

Benzyl piperazine (BZP) dihydrochloride

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2 · 2HCl
CAS Number:
Molecular Weight:
249.18
UNSPSC Code:
41116107
Product NMID905 is not currently sold in your country. Contact Technical Service

grade

certified reference material

form

solid

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

room temp

InChI

1S/C11H16N2/c1-2-4-11(5-3-1)10-13-8-6-12-7-9-13/h1-5,12H,6-10H2

InChI key

IQXXEPZFOOTTBA-UHFFFAOYSA-N

General description

A reference material suitable for LC/MS or GC/MS amphetamine testing methods in clinical toxicology, forensic analysis, or urine drug testing. Benzyl piperazine, or BZP, is an amphetamine analog used illicitly for its euphoriant and stimulant effects.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service