All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
UNSPSC Code:
41116107
manufacturer/tradename
NMI Australia
format
neat
storage temp.
room temp
InChI
1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2
InChI key
SATCULPHIDQDRE-UHFFFAOYSA-N
signalword
Warning
hcodes
pcodes
Hazard Classifications
Skin Sens. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
150.1 °F
flash_point_c
65.62 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service