Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID785B

Hydromorphone hydrochloride

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3 · HCl
CAS Number:
Molecular Weight:
321.80
UNSPSC Code:
41116107

manufacturer/tradename

NMI Australia

format

neat

storage temp.

room temp

InChI

1S/C17H19NO3.ClH/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;/h2,4,10-11,16,19H,3,5-8H2,1H3;1H/t10-,11+,16-,17-;/m0./s1

InChI key

XHILEZUETWRSHC-NRGUFEMZSA-N

General description

Reference material suitable for use as a starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research. Hydromorphone, marketed under brand names including Palladone and Dilaudid, is a very potent opioid analgesic. The drug is often used medicinally as an alternative to morphine for treatment of pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service