Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID709

d3-Morphine

NMI Australia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16D3NO3
CAS Number:
Molecular Weight:
288.36
UNSPSC Code:
41116107

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C17H19NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2-5,10-11,13,16,19-20H,6-8H2,1H3/t10-,11+,13-,16-,17-/m0/s1/i1D3

InChI key

BQJCRHHNABKAKU-LLQNGXJRSA-N

General description

This reference material is suitable for quantiation of morphine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, or isotope dilution methods. Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin, Kadian, Oramorph, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service