Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NMID615B

6ß-Hydroxy-oral turinabol (available to WADA laboratories only)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27ClO3
CAS Number:
Molecular Weight:
350.88
UNSPSC Code:
41116107

grade

certified reference material

agency

NMI Australia certified reference material

form

solid

packaging

pkg of ~1 mg

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service