NMID567
2α-Methyl-5α-androstan-3α-ol-17-one
NMI Australia
About This Item
Recommended Products
agency
NMI Australia certified reference material
manufacturer/tradename
NMI Australia
format
neat
storage temp.
2-8°C
InChI
1S/C20H32O2/c1-12-11-20(3)13(10-17(12)21)4-5-14-15-6-7-18(22)19(15,2)9-8-16(14)20/h12-17,21H,4-11H2,1-3H3/t12-,13+,14+,15+,16+,17+,19+,20+/m1/s1
InChI key
FDGQIVOIHOIMNK-URBDGJAYSA-N
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service