Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-143

Supelco

Norhydrocodone-D3 hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$373.00

$373.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$373.00

About This Item

Empirical Formula (Hill Notation):
C17D3H16NO3 · HCl
Molecular Weight:
324.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$373.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.[2H]C([2H])([2H])Oc1ccc2C[C@H]3NCC[C@@]45[C@H]3CCC(=O)[C@@H]4Oc1c25

InChI

1S/C17H19NO3.ClH/c1-20-13-5-2-9-8-11-10-3-4-12(19)16-17(10,6-7-18-11)14(9)15(13)21-16;/h2,5,10-11,16,18H,3-4,6-8H2,1H3;1H/t10-,11+,16-,17-;/m0./s1/i1D3;

InChI key

IQVZQPJFFRBEHK-PPYOUCNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard applicable for quantitation of hydrocodone levels in urine, serum or plasma by LC/MS or GC/MS for pain prescription monitoring, urine drug testing, clinical toxicology, forensic analysis or pharmaceutical research. Norhydrocodone is a primary urinary metabolite of the analgesic hydrocodone. Hydrocodone, a semi-synthetic opioid derived from codeine and thebaine, is used as a painkiller and as an antitussive.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service