Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-125

Supelco

Nortetrazepam solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15ClN2O
CAS Number:
Molecular Weight:
274.75
MDL number:
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C15H15ClN2O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h4,6-8H,1-3,5,9H2,(H,18,19)

InChI key

FDRMSENAXZDFTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nortetrazepam is a major metabolite of tetrazepam. A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for nortetrazepam LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service