Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-092

Supelco

4-(Methylnitrosamine)-1-(3-pyridyl)-1-butanone-13C6 (NNK-13C6) solution

100 μg/mL in methanol (), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C413C6H13N3O2
CAS Number:
Molecular Weight:
213.19
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (
)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Also known as nicotine-derived nitrosamine ketone, NNK is a potent carcinogen and biomarker of exposure to cigarette smoke. NNK, produced upon the curing of tobacco, is found in a variety of tobacco products including chewing tobacco, snuff, cigarettes, and cigars. This stable-labeled internal standard is suitable for use in LC-MS/MS or GC/MS applications in nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service