Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-013

Supelco

Noroxymorphone hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO4·HCl
CAS Number:
Molecular Weight:
323.77
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

Cl.Oc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O

InChI

1S/C16H17NO4.ClH/c18-9-2-1-8-7-11-16(20)4-3-10(19)14-15(16,5-6-17-11)12(8)13(9)21-14;/h1-2,11,14,17-18,20H,3-7H2;1H/t11-,14+,15+,16-;/m1./s1

InChI key

QMFVIJMHPXUVOL-RCGDHTHDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution of noroxymorphone suitable for use in a variety of GC/MS or LC/MS applications from pain prescription monitoring, clinical toxicology, and pharmaceutical research to forensic analysis and urine drug testing. Noroxymorphone is a major metabolite of oxymorphone in urine. Oxymorphone, sold under trade names such as Opana®, Numorphan®, and Numorphone, is an opiate analgesic indicated for the relief of moderate to severe pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Numorphan is a registered trademark of Endo Pharmaceuticals Inc.
Opana is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service