Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-195

Supelco

Metformin Impurity Mixture hydrochloride solution

50 μg/mL in methanol (each component), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

50 μg/mL in methanol (each component)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

multi-component solution

storage temp.

−20°C

General description

Metformin, marketed under the trade name Glucophage among others, is a first-line medication for the treatment of type 2 diabetes and polycystic ovary syndrome. This solution mix Certified Reference Material contains metformin along with multiple impurities produced during pharmaceutical manufacturing of metformin - 1-methylbiguanide HCl (Metformin Related Compound B); melamine; and N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine (Metformin Related Compound C). Used with M-196-1ML Cyanoguanidine (Metformin Related Compound A), this certified solution standard is suitable for use in metformin monograph testing methods by HPLC in testing applications ranging from API release testing to pharmaceutical research.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Metformin Related Compound B, Pharmaceutical Secondary Standard; Certified Reference Material
    Metformin Related Compound C, Pharmaceutical Secondary Standard; Certified Reference Material
    Melamine, Pharmaceutical Secondary Standard; Certified Reference Material
    Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service