Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-175

Supelco

MDPBP-D8 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11D8NO3 · HCl
CAS Number:
Molecular Weight:
305.83
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

Storage temp.

−20°C

SMILES string

CCC(N1C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H])C(C2=CC(OCO3)=C3C=C2)=O.[H]Cl

InChI

1S/C15H19NO3.ClH/c1-2-12(16-7-3-4-8-16)15(17)11-5-6-13-14(9-11)19-10-18-13;/h5-6,9,12H,2-4,7-8,10H2,1H3;1H/i3D2,4D2,7D2,8D2;

Inchi Key

ATCKCJHEOJFKJW-SOLWPGNYSA-N

General description

A novel designer drug first developed in the 1960′s as a research chemical, MDPBP emerged as a legal high in Europe in 2009. This synthetic cathinone and bath salt is often sold in the US and other countries under the name "NRG-1" as a mixture with other substituted cathinone derivatives such as flephedrone, pentylone, and MDPV. This Certified Spiking Solution® is suitable as an internal standar for MDPBPP testing methods by GC/MS or LC/MS in forensic analysis, clinical toxicology, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service