Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)
concentration
100 μg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
General description
25I-NBOMe is a second generation 2C amine derived from attaching an NBOMe (2-methoxybenzyl) group to the amine group of 2C-I. Sold under street names such as N-bomb, Smiles and 25I, this designer drug produces vivid visual hallucinations similar to those of LSD. This internal standard is suitable for quantitation of 25I-NBOMe in LC/MS or GC/MS testing methods for forensic analysis, urine drug testing, or clinical toxicology applications.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.