Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I-014

Supelco

(±)-5-Iodo-2-aminoindane hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10NI · HCl
Molecular Weight:
295.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

NC(C1)CC2=C1C=CC(I)=C2.Cl

InChI

1S/C9H10IN.ClH/c10-8-2-1-6-4-9(11)5-7(6)3-8;/h1-3,9H,4-5,11H2;1H

InChI key

KXPCULLKBZKNFC-UHFFFAOYSA-N

General description

This certified Snap-N-Spike® solution is suitable for use in designer drug testing methods by LC/MS or GC/MS in clinical toxicology, urine drug testing, or forensic analysis applications. 5-Iodo-2-aminoindane, also known as 5-IAI, is an MDMA analog similar in structure to the designer drug MDAI. The drug acts as a releasing agent of serotonin, norepinephrine, and dopamine to produce psychedelic effects similar to those of MDMA.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service