Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-136

Supelco

4-Hydroxyamphetamine hydrochloride solution

1.0 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO · HCl
CAS Number:
Molecular Weight:
187.67
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.NC(Cc1ccc(cc1)O)C

InChI

1S/C9H13NO.ClH/c1-7(10)6-8-2-4-9(11)5-3-8;/h2-5,7,11H,6,10H2,1H3;1H

InChI key

MYTMYIPUXIMVQW-UHFFFAOYSA-N

General description

4-Hydroxyamphetamine, also known as 4-HA, para-hydroxyamphetamine, and α-methyltyramine, is a drug that stimulates the sympathetic nervous system. It is a major metabolite of amphetamine and certain substituted amphetamines. This Certified Spiking Solution® is applicable for use in urine drug testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service