Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-117

Supelco

4-Hydroxyduloxetine acetate solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO2S · C2H4O2
Molecular Weight:
373.47
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

General description

4-Hydroxyduloxetine is a major metabolite of duloxetine. Duloxetine, marketed in the US under the trade name Cymbalta®, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis. This Certified Spiking Solution® is applicable for use in applications such as clinical toxicology, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Cymbalta is a registered trademark of Eli Lilly and Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service