Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-116

Supelco

3-Hydroxyflubromazepam solution

1.0 mg/mL (10% DMSO in Acetonitrile), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O2FBr
CAS Number:
Molecular Weight:
349.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (10% DMSO in Acetonitrile)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

BrC1=CC=C2C(C(C3=CC=CC=C3F)=NC(O)C(N2)=O)=C1

InChI

1S/C15H10BrFN2O2/c16-8-5-6-12-10(7-8)13(19-15(21)14(20)18-12)9-3-1-2-4-11(9)17/h1-7,15,21H,(H,18,20)

InChI key

LAAYFWALJDHMAB-UHFFFAOYSA-N

General description

A primary urinary metabolite of flubromazepam, 3-hydroxyflubromazepam is a synthetic benzodiazepine and structural analog of phenazepam sold on the illicit market as a novel designer drug. This certified solution standard is suitable as starting material in calibrators and controls for GC/MS or LC/MS benzodiazepine testing applications in clinical toxicology, urine drug testing or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service