Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-114

Supelco

3-Hydroxyphenazepam-D4 solution

100 μg/mL in acetonitrile, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H6D4BrClN2O2
Molecular Weight:
369.63
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

BrC1=CC(C(C2=C([2H])C([2H])=C([2H])C([2H])=C2Cl)=NC3O)=C(C=C1)NC3=O

InChI

1S/C15H10BrClN2O2/c16-8-5-6-12-10(7-8)13(19-15(21)14(20)18-12)9-3-1-2-4-11(9)17/h1-7,15,21H,(H,18,20)/i1D,2D,3D,4D

InChI key

KRJKJUWAZOWXNV-RHQRLBAQSA-N

General description

An internal standard suitable for quantitation of 3-Hydroxyphenazepam in mass-spectrometry based testing methods for forensic analysis, urine drug testing, or clinical toxicology applications. 3-Hydroxyphenazepam is an active metabolite of the designer benzodiazepine and novel psychoactive substance (NPS), phenazepam, which emerged in online drug markets in late 2011. Despite being a controlled substance in Europe, North America, and many countries in Asia, phenazepam has been discovered in LSD-type blotters as well as some synthetic cannabinoid blends.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Moran et al.
Oncogene, 27(42), 5567-5577 (2008-05-28)
P53 wild-type and p53-null or mutant cells undergo a G(2)-phase cell-cycle arrest in response to ionizing radiation (IR). In this study we examined the effect of heat-shock protein 90 (HSP90) inhibitor, geldanamycin (GA), on IR-induced G(2) arrest in human colon
Joana M P Desterro et al.
Journal of cell science, 116(Pt 9), 1805-1818 (2003-04-01)
ADAR1 and ADAR2 are editing enzymes that deaminate adenosine to inosine in long double stranded RNA duplexes and specific pre-mRNA transcripts. Here, we show that full-length and N-terminally truncated forms of ADAR1 are simultaneously expressed in HeLa and COS7 cells
S Pust et al.
Oncogene, 32(29), 3443-3451 (2012-08-08)
Amplification and overexpression of the receptor tyrosine kinase ErbB2 occur in up to 30% of human breast cancers, and high ErbB2 levels are correlated with poor prognosis for breast cancer patients. In contrast to the epithelial growth factor receptor (ErbB1)
O Mortusewicz et al.
Oncogene, 35(6), 761-770 (2015-05-12)
The transcriptional cofactor PC4 is an ancient single-strand DNA (ssDNA)-binding protein that has a homologue in bacteriophage T5 where it is likely the elusive replicative ssDNA-binding protein. We hypothesize that human PC4 has retained functions in ssDNA binding to stabilize
Rachael Barry et al.
Nature communications, 9(1), 3001-3001 (2018-08-03)
The NLRP3 inflammasome responds to infection and tissue damage, and rapidly escalates the intensity of inflammation by activating interleukin (IL)-1β, IL-18 and cell death by pyroptosis. How the NLRP3 inflammasome is negatively regulated is poorly understood. Here we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service