Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-022

Supelco

Guaifenesin Impurity Mixture solution

0.5-1.0 mg/mL in acetonitrile (each analyte), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
Molecular Weight:
124.14
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Guaiacol, 0.5 mg/mL
Guaifenesin, 1 mg/mL
Guaifenesin β-isomer, 0.5 mg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

0.5-1.0 mg/mL in acetonitrile (each analyte)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

multi-component solution

storage temp.

−20°C

General description

Guaifenesin, a glyceryl ether of guaiacol, is a key constituent in cold and cough formulations. It belongs to the group of expectorant medicines because of its ability to ease the discomfort caused due to cough by causing an increase in the sputum volume and a simultaneous decrease in its viscosity. Its applications include short term relief in the breathing passageway in various conditions such as allergy, cold, and upper respiratory tract infections. It has also been proposed to be effective in the management of pain in various conditions such as prostatitis, seminal vesiculitis, benign prostatic hypertrophy, prostatic abscess, bladder neck hypertrophy, dysmenorrhea, and urinary tract infection.[1][2][3]

Application

Guaifenesin Impurity Mixture solution may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Guaifenesin, certified reference material, pharmaceutical secondary standard
    Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
    Guaifenesin Impurity B, Pharmaceutical Secondary Standard; Certified Reference Material

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guaifenesin
USP42-NF37
United States Pharmacopeia/National Formulary, 41(5), 2121-2121 (2018)
Guaifenesin as a treatment for primary dysmenorrhea
Marsden JS, et al
Journal of the American Board of Family Medicine : JABFM, 17(4), 240-246 (2004)
Effect of guaifenesin on cough reflex sensitivity
Dicpinigaitis VP, et al.
Chest, 124(6), 2178-2181 (2003)
Voltammetric assay of Guaifenesin in pharmaceutical formulation
Tapsoba I, et al.
Journal of Pharmaceutical and Biomedical Analysis, 38(1), 162-165 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service