Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-014

Supelco

Ginkgo Biloba Flavonoids Mix

100 μg/mL each component in methanol, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O7
Molecular Weight:
316.26
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Isorhamnetin
Kaempferol
Quercetin

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL each component in methanol

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

Ginkgo biloba extract is a mixture of biologically active secondary metabolites. The main constituents are bilobalide (BB), ginkgolide A (GA), ginkgolide B (GB) and ginkgolide C (GC).[1][2] Ginkgo biloba extracts include around 300 compounds, with major constituents being flavonoids and terpene lactones. Flavonoids make up to 24% of the total extract.[3]

Biochem/physiol Actions

Ginkgo biloba terpene lactone constitute a major part of Ginkgo Biloba extract (GBE), which is used as a phytomedicine, and is used for multiple disorders including CNS (central nervous system), circulatory, cardiovascular, respiratory and renal diseases.[3] Extracts from the leaves of Gingko plant are thought to have anti-stress, anti-cancer, neuroprotective, cardioprotective and memory enhancing properties. Flavonoids are thought play a complementary role in these effects of GBE.[4]

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mahadevan et al.
Journal of food science, 73(1), R14-R19 (2008-01-24)
The new age of nutraceuticals is now embracing the centuries old herbal extract of Ginkgo biloba (Mantissa Plantarum Altera, 1771, Ginkgoceae). The standardized preparation of the Ginkgo leaf extract (EGb 761) contained 2 main bioactive constituents, flavonoid glycosides (24%) and
Yong Joo Park et al.
Asian Pacific journal of cancer prevention : APJCP, 16(15), 6317-6325 (2015-10-06)
Ginkgo biloba extract (GBE) is a popular phytomedicine and has been used for disorders of the central nervous system, cardiovascular, renal, respiratory, and circulatory diseases. Although GBE is a complex mixture of over 300 compounds, its major components are 24%
Mostafa K Mesbah et al.
Farmaco (Societa chimica italiana : 1989), 60(6-7), 583-590 (2005-06-07)
The biologically active secondary metabolites of Ginkgo biloba extract EGb 761 in phytopharmaceuticals were analyzed using two simple, rapid, accurate and sensitive HPLC methods. The proposed methods were successfully applied in the determination of terpenes and flavonoids in four phytopharmaceutical
Ping Huang et al.
Journal of pharmaceutical and biomedical analysis, 100, 138-144 (2014-08-29)
The ginkgo terpene lactones (GTL), mainly including bilobalide (BB), ginkgolide A (GA), ginkgolide B (GB) and ginkgolide C (GC) possess different biological activities such as peripheral vasoregulation, platelet-activating factor (PAF) receptor antagonism, neuroprotective properties and prevention of membrane damage caused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service