Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-058

Supelco

4-Fluoroisobutyryl fentanyl-13C6 solution

100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C1713C6H29FN2O
Molecular Weight:
374.44
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

This stable-labeled internal standard is suitable for quantitation of 4-fluoroisobutyryl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods. 4-Fluoroisobutyryl fentanyl is an opioid analgesic analog of fentanyl and has been sold as a designer drug on the illicit drug market.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service