Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-042

Supelco

(±)-3-Fluorophenmetrazine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14FNO · HCl
CAS Number:
Molecular Weight:
231.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

FC1=CC=CC(C2C(C)NCCO2)=C1.[H]Cl

InChI

1S/C11H14FNO.ClH/c1-8-11(14-6-5-13-8)9-3-2-4-10(12)7-9;/h2-4,7-8,11,13H,5-6H2,1H3;1H

InChI key

MHJBXKFJWSBWRE-UHFFFAOYSA-N

General description

(±)-3-Fluorophenmetrazine (3-FPM) is the 3′-fluoro analogue of phenmetrazine, a withdrawn anorectic drug with amphetamine-like effects. 3-FPM has been sold online as a designer stimulant and reportedly exerts strong euphoric effects. This certified reference standard is suitable for use in a wide variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing and pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service