Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-027

Supelco

(±)-4-Fluoroephedrine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14FNO · HCl
CAS Number:
Molecular Weight:
219.68
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

CC(NC)C(O)C1=CC=C(F)C=C1.Cl

InChI

1S/C10H14FNO.ClH/c1-7(12-2)10(13)8-3-5-9(11)6-4-8;/h3-7,10,12-13H,1-2H3;1H

InChI key

FENYPFLMDTWUKZ-UHFFFAOYSA-N

General description

A primary urinary metabolite and synthetic cathinone analog of Flephedrone. Flephedrone, also known as 4-Fluoromethcathinone or 4-FMC, is a bath salt cathinone and stimulant widely found in numerous bath salt blends across the US, Europe, and Asia. This certified solution standard is suitable as starting material for calibrators and controls in flephedrone testing methods by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service