Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-022

Supelco

(±)-4-Fluoroamphetamine-D5 (side chain) hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7D5FN · HCl
Molecular Weight:
194.69
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

NC([2H])(C([2H])([2H])[2H])C([2H])C1=CC=C(F)C=C1.[H]Cl

InChI

1S/C9H12FN.ClH/c1-7(11)6-8-2-4-9(10)5-3-8;/h2-5,7H,6,11H2,1H3;1H/i1D3,6D,7D;

InChI key

GKWYMWZWSCKSMT-NINFPRRKSA-N

General description

A fluoro analog of amphetamine, 4-Fluoroamphetamine is sold in the illicit market as a club/designer drug under street names including R2D2, Flux and Flits. The drug produces stimulant and possibly empathogenic or psychoactive effects. This internal standard is suitable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service