Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERT-091

Supelco

Petroleum Hydrocarbon Surrogate Solution

 μg/mL in methylene chloride (varied conc.), ampule of 5 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

composition

o-Terphenyl, 10000 μg/mL
α,α,α-Trifluorotoluene, 20000 μg/mL

packaging

ampule of 5 mL

manufacturer/tradename

Cerilliant®

concentration

 μg/mL in methylene chloride (varied conc.)

application(s)

environmental
petroleum

format

single component solution

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    α,α,α-Trifluorotoluene 20000 μg/mL
    o-Terphenyl 10000 μg/mL

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service