Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERM-041

6-Methylchrysene

vial of 10 mg, analytical standard, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H14
CAS Number:
Molecular Weight:
242.31
UNSPSC Code:
41116107

grade

analytical standard

packaging

vial of 10 mg

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

Storage temp.

room temp

SMILES string

Cc1cc2c3ccccc3ccc2c4ccccc14

InChI

1S/C19H14/c1-13-12-19-16-8-3-2-6-14(16)10-11-18(19)17-9-5-4-7-15(13)17/h2-12H,1H3

Inchi Key

ASVDRLYVNFOSCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Amin et al.
Cancer research, 45(12 Pt 1), 6406-6442 (1985-12-01)
We compared the metabolic activation in mouse skin of the weak carcinogen 6-methylchrysene, which lacks a bay region methyl group, and the strong carcinogen 5-methylchrysene, which has a bay region methyl group. Metabolites of 6-methyl-chrysene were prepared using liver homogenates
S S Hecht et al.
Cancer research, 47(20), 5310-5315 (1987-10-15)
The stereochemistry of diol epoxide formation in mouse epidermis upon topical application of [3H]-1R,2R-dihydroxy-1,2-dihydro-5-methylchrysene ([3H]-5-MeC-1R,2R-diol) and [3H]-6-MeC-1R,2R-diol, and the tumorigenicity in mouse skin and in newborn mice of the R,S,S,R and S,R,R,S enantiomers of 1,2-dihydroxy-3,4-epoxy-1,2,3,4-tetrahydro-5-methylchrysene (5-MeC-1,2-diol-3,4-epoxide), 5-MeC-7,8-diol-9,10-epoxide, and 6-MeC-1,2-diol-3,4-epoxide were
S R Myers et al.
Chemico-biological interactions, 77(2), 203-221 (1991-01-01)
The polynuclear aromatic hydrocarbon chrysene undergoes a bioalkylation substitution reaction in vitro, in rat liver cytosol preparations, and in vivo, in rat dorsal subcutaneous tissue to yield 6-methylchrysene as a metabolite. In addition, both 5-methyl- and 6-methylchrysene were found to
S Amin et al.
Cancer research, 47(14), 3613-3617 (1987-07-15)
The stereoselectivity of mouse skin metabolic activation to dihydrodiols of the strong carcinogen 5-methylchrysene (5-MeC) and the weak carcinogen 6-methylchrysene (6-MeC) was investigated. Synthetic 1,2-dihydro-1,2-dihydroxy-5-methylchrysene (5-MeC-1,2-diol), 5-MeC-7,8-diol, and 6-MeC-1,2-diol were resolved into their R,R- and S,S-enantiomers by chiral stationary phase
W Koehl et al.
Cancer research, 56(2), 316-324 (1996-01-15)
The metabolism of environmentally occurring methylated polynuclear aromatic hydrocarbons by human cytochrome P450 (P450) enzymes has not been examined previously. We compared the metabolism of the tobacco smoke constituents 5-methylchrysene (5-MeC), a strong carcinogen, and 6-MeC, a weak carcinogen, in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service