Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERH-050S

Supelco

HMX solution

10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N8O8
CAS Number:
Molecular Weight:
296.16
MDL number:
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 5 mL

manufacturer/tradename

Cerilliant®

concentration

10 mg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])N1CN([N+]([O-])=O)CN([N+]([O-])=O)CN([N+]([O-])=O)C1

InChI

1S/C4H8N8O8/c13-9(14)5-1-6(10(15)16)3-8(12(19)20)4-7(2-5)11(17)18/h1-4H2

InChI key

UZGLIIJVICEWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudeep Perumbakkam et al.
Research in microbiology, 163(8), 567-575 (2012-08-21)
Explosives such as octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) provide a challenge in terms of bioremediation. In the present study, sheep rumen was studied for its potential to detoxify HMX using analytical chemistry and molecular microbial ecology tools. Results indicated significant loss (p <
Guilherme R Lotufo
Ecotoxicology and environmental safety, 74(3), 301-306 (2011-01-25)
Sheepshead minnows (Cyprinodon variegatus) were exposed to radiolabeled isotopes of the explosives 2,4,6-trinitrotoluene (TNT), exahydro-1,3,5-trinitro-1,3,5-triazine (commonly known as RDX), and octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (commonly known as HMX), yielding the bioconcentration factors (BCF) of 3.3, 0.7, and 0.1 L kg(-1), respectively. For TNT
Thomas A Douglas et al.
Environmental toxicology and chemistry, 30(2), 345-353 (2010-11-03)
Potentially toxic nitroaromatic and nitramine compounds are introduced onto soils during detonation of explosives. The present study was conducted to investigate the desorption and transformation of explosive compounds loaded onto three soils through controlled detonation. The soils were proximally detonated
Chun-jiang An et al.
Journal of hazardous materials, 179(1-3), 526-532 (2010-04-03)
The performance of mesophilic anaerobic granules to degrade octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) was investigated under various conditions. The results of batch experiments showed that anaerobic granules were capable of removing HMX from aqueous solution with high efficiency. Both biotic and abiotic mechanisms
S T McMurry et al.
Ecotoxicology (London, England), 21(2), 304-314 (2011-09-29)
Environmental contamination by energetic compounds is an increasing international concern, although little is known of their accumulation in and affect on wildlife. Reptiles are often good models for contaminants studies due to natural history traits that increase their potential for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service