Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERC-012

Supelco

p-Chlorophenyl methyl sulfone solution

vial of 100 mg, analytical standard, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClO2S
CAS Number:
Molecular Weight:
190.65
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

vial of 100 mg

manufacturer/tradename

Cerilliant®

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C7H7ClO2S/c1-11(9,10)7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

LMCOQDVJBWVNNI-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of 4-chlorophenyl methyl sulfide and its sulfone analog in cattle and sheep.
D D Oehler et al.
Archives of environmental contamination and toxicology, 12(2), 227-233 (1983-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service