Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERC-003

Supelco

cis-Chlordane

vial of 25 mg, analytical standard, (α isomer), Cerilliant®

Synonym(s):

α-Chlordane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Cl8
CAS Number:
Molecular Weight:
409.78
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

vial of 25 mg

manufacturer/tradename

Cerilliant®

application(s)

agriculture
environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

Cl[C@H]1CC2C([C@H]1Cl)[C@@]3(Cl)C(Cl)=C(Cl)[C@]2(Cl)C3(Cl)Cl

InChI

1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2?,3-,4?,5-,8-,9+/m0/s1

InChI key

BIWJNBZANLAXMG-MRAITHJBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianqiao Xu et al.
Environmental science & technology, 48(14), 8012-8020 (2014-06-17)
Bioconcentration factors (BCFs) measured in the laboratory are important for characterizing the bioaccumulative properties of chemicals entering the environment, especially the potential persistent organic pollutants (POPs), which can pose serious adverse effects on ecosystem and human health. Traditional lethal analysis
Shuqin Liu et al.
Analytica chimica acta, 878, 109-117 (2015-05-24)
Mesoporous TiO2 nanoparticles were synthesized with the hydrothermal method and characterized by powder X-ray diffraction (PXRD) and transmission electron microscope (TEM). Then a superior solid-phase microextraction (SPME) fiber was fabricated by sequentially coating the stainless steel fiber with silicone sealant
Yunbin Jiang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 983-984, 47-54 (2015-01-27)
A rapid and accurate multi-residue method was developed for simultaneously monitoring and determining 34 kinds of pesticides by GC-EI/MS/MS, successfully applied in Qianjinzhidai pills. Precision, repeatability, stability, limit of detection (LOD), limit of quantification (LOQ), linearity range and accuracy tests
Jing Zhan et al.
Food chemistry, 194, 319-324 (2015-10-17)
A simple, rapid and effective method was developed based on matrix solid-phase dispersion (MSPD) for the determination of organochlorine pollutants including sixteen organochlorine pesticides (OCPs) and seven polychlorinated biphenyls (PCBs) in oil seeds (peanuts and soybeans). Among the organochlorine pollutants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service