Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E-500

Supelco

Estrogens CRM in Serum solution

0 pg/mL (Sigmatrix Ultra), certified reference material, vial of 3 mL (Blank)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
Molecular Weight:
270.37
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

17ß-Estradiol, 0 pg/mL (each analyte)
Estrone, 0 pg/mL (each analyte)

packaging

vial of 3 mL (Blank)

manufacturer/tradename

Cerilliant®

concentration

0 pg/mL (Sigmatrix Ultra)

application(s)

clinical testing

format

multi-component solution

storage temp.

−20°C

General description

Certified Reference Material of estrone and estradiol free serum for clinical research applications by mass spectrometry. Estrogens blank is ready to be used as blank serum matrix for linearity standards or for calibration verification in LC-MS/MS estrogens testing methods. Separate options for individual levels of estrone and estradiol in serum at concentrations ranging from 10 to 1,000 pg/mL for each analyte and 13C- or deuterium labeled internal standards of estrone and estradiol are also available.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service