E-085
(±)-N-Ethylcathinone Ephedrine Metabolite-D5 hydrochloride solution
100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
About This Item
grade
certified reference material
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
Storage temp.
−20°C
SMILES string
CC(C(O)C1=CC=CC=C1)NC([2H])([2H])C([2H])([2H])[2H].Cl
InChI
1S/C11H17NO.ClH/c1-3-12-9(2)11(13)10-7-5-4-6-8-10;/h4-9,11-13H,3H2,1-2H3;1H/i1D3,3D2;
Inchi Key
DHNCTQIFOGRFJF-IYSLTCQOSA-N
General description
Legal Information
related product
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service