Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E-019

Supelco

Ecgonine ethyl ester solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CCOC(=O)[C@H]1[C@@H](O)C[C@@H]2CC[C@H]1N2C

InChI

1S/C11H19NO3/c1-3-15-11(14)10-8-5-4-7(12(8)2)6-9(10)13/h7-10,13H,3-6H2,1-2H3/t7-,8+,9-,10+/m0/s1

InChI key

ABDMBNAKYBUEEX-QCLAVDOMSA-N

General description

A certified solution standard suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing to forensic analysis and clinical toxicology. Ecgonine Ethyl Ester is a unique biomarker that indicates the simultaneous consumption of cocaine and alcohol. Ecgonine Ethyl Ester is a metabolite found in blood obtained by cocaine hydrolysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Giroud et al.
Schweizerische Rundschau fur Medizin Praxis = Revue suisse de medecine Praxis, 82(15), 441-446 (1993-04-13)
Cocaine is a powerful reinforcer that has become a popular drug of abuse. Cocaine inhibits the reuptake of dopamine into presynaptic terminals, resulting in an overstimulation of the dopaminergic system. When cocaine and alcohol are taken together, cocaine is partially
Loralie J Langman et al.
Journal of analytical toxicology, 33(8), 447-455 (2009-10-31)
Cocaine (COC) is a potent CNS stimulant that is metabolized to benzoylecgonine (BE) and further metabolized to minor metabolites such as m-hydroxybenzoylecgonine (m-HOBE). COC is also metabolized to norcocaine (NC). Cocaethylene (CE) is formed when cocaine and ethyl alcohol are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service