Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E-012

Supelco

EDDP perchlorate solution

100 μg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Methadone metabolite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClNO4
CAS Number:
Molecular Weight:
377.86
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as pyrolinium)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

Storage temp.

−20°C

SMILES string

C[N+]1=C(CC)C(C2=CC=CC=C2)(C3=CC=CC=C3)CC1C.O=Cl(=O)([O-])=O

InChI

1S/C20H24N.ClHO4/c1-4-19-20(15-16(2)21(19)3,17-11-7-5-8-12-17)18-13-9-6-10-14-18;2-1(3,4)5/h5-14,16H,4,15H2,1-3H3;(H,2,3,4,5)/q+1;/p-1

Inchi Key

FICCBMGBBWWZRF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution of the major urinary metabolite of methadone. Methadone, sold under various trade names such as Symoron, Amidone or Methadose®, is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. Suitable uses for this reference standard include as starting material in preparation of calibrators and controls for LC/MS or GC/MS methods in methadone testing for applications in clinical toxicology or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Methadose is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Marie Dollerup Holm et al.
Journal of analytical toxicology, 36(7), 487-496 (2012-07-11)
We developed a method based on liquid chromatography coupled with tandem mass spectrometry to quantify individual enantiomers of methadone and its primary metabolite, R/S-2-ethyl-1,5-dimethyl-3,3-diphenylpyrrolinium (EDDP), in postmortem blood and brain tissue. Samples were prepared with a Tecan Evo robotic system.
Maksymilian Kulza et al.
Przeglad lekarski, 67(10), 925-928 (2010-01-01)
Methadone maintenance therapy is one of the most popular treatments for opiate addiction. The easiest and the most reliable way to monitor the accuracy of the therapy is determination of methadone and its metabolites in biological material. The aim of
Ricarda Jantos et al.
Forensic science international, 226(1-3), 254-260 (2013-02-21)
Body fluids and tissues in 16 methadone (MTD)-associated fatalities were investigated to find out whether analysis of MTD and its metabolite 2-ethylidine-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) following enantioselective separation of both compounds may assist in the interpretation of MTD findings. Individual case histories
Cristiana Gambelunghe et al.
Annals of clinical and laboratory science, 43(1), 22-30 (2013-03-07)
It may be advantageous to use sweat, rather than blood or urine, to monitor individuals' drug exposure for the purposes of drug treatment programs, employment initiatives, and forensic investigations. Forty-eight patients receiving methadone at the Public Service for the Treatment
Mei-Han Huang et al.
Toxicology, 280(1-2), 1-9 (2010-11-26)
Methadone (Mtd) is a widely used opioid drug associated with the side effect of hyperprolactinemia. The mechanism of how Mtd induces prolactin secretion remains unclear. The effects of Mtd and its two main metabolites (EDDP: (±)-2-ethyl-1,5-dimethyl-3,3-diphenylpyrrolinium percholarate and EMDP: 2-ethyl-5-methyl-3,3-dipnehyl-1-pyrroline)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service