Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-183

Supelco

Diphenhydramine Impurity C solution

500 μg/mL (Acetonitrile:0.05% aqueous phosphoric acid (70:30) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20BrNO · HCl
Molecular Weight:
370.71
MDL number:
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL (Acetonitrile:0.05% aqueous phosphoric acid (70:30) (v/v))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

Storage temp.

−20°C

General description

Diphenhydramine is an active ingredient that is used to treat symptoms caused by histamines suffered because of the common cold or allergies. This certified solution standard contains Diphenhydramine Impurity C. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

42.8 °F - closed cup

flash_point_c

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service