Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-173

Supelco

Dothiepin HCl (cis/trans) solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NS · HCl
CAS Number:
Molecular Weight:
331.90
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

S1Cc2c(cccc2)\C(=C\CCN(C)C)\c3c1cccc3.[Cl-].[H+]

InChI

1S/C19H21NS.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11-;

InChI key

XUPZAARQDNSRJB-CULRIWENSA-N

General description

Dothiepin, also known as Dosulepin and marketed as Prothiaden among others, is a tricyclic antidepressant (TCA) which is used in the treatment of depression. Dothiepin also has antihistamine, antiadrenergic, antiserotonergic, anticholinergic, and sodium channel-blocking effects. Due to its high toxicity and risk of overdose dothiepin is assayed in therapeutic drug monitoring to ensure patients remain within the drug′s therapeutic range. This certified Snap-N-Spike® solution is suitable for LC-MS/MS applications including therapeutic drug monitoring, clinical toxicology, urine drug testing or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service