Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-170

Supelco

Dexmethylphenidate hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO2 · HCl
CAS Number:
Molecular Weight:
269.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1 mg/mL in methanol (as free base)

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C14H19NO2.ClH/c1-17-14(16)13(11-7-3-2-4-8-11)12-9-5-6-10-15-12;/h2-4,7-8,12-13,15H,5-6,9-10H2,1H3;1H/t12-,13-;/m1./s1

InChI key

JUMYIBMBTDDLNG-OJERSXHUSA-N

General description

Dexmethylphenidate, sold under the brand name Focalin among others, is the active dextrorotatory enantiomer of methylphenidate. Dexmethylphenidate is a central nervous system (CNS) stimulant prescribed for treatment of attention deficit hyperactivity disorder (ADHD). This Certified Spiking Solution®; is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ni Huang et al.
Nature medicine, 27(5), 892-903 (2021-03-27)
Despite signs of infection-including taste loss, dry mouth and mucosal lesions such as ulcerations, enanthema and macules-the involvement of the oral cavity in coronavirus disease 2019 (COVID-19) is poorly understood. To address this, we generated and analyzed two single-cell RNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service