Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-084

Supelco

Desmethylene Paroxetine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FNO3 · HCl
CAS Number:
Molecular Weight:
353.82
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

OC(C=C1)=C(O)C=C1OC[C@@H]2CNCC[C@H]2C3=CC=C(F)C=C3.Cl

InChI

1S/C18H20FNO3.ClH/c19-14-3-1-12(2-4-14)16-7-8-20-10-13(16)11-23-15-5-6-17(21)18(22)9-15;/h1-6,9,13,16,20-22H,7-8,10-11H2;1H/t13-,16-;/m0./s1

InChI key

GIPUDSSHAJOHQK-LINSIKMZSA-N

General description

Desmethylene paroxetine is a major urinary metabolite of the SSRI antidepressant paroxetine. Paroxetine, marketed under trade names such as Paxil® or Aropax, is used to treat many conditions in adults from major depression and obsessive-compulsive disorder to several anxiety disorders. Suitable uses for this certified solution standard include as a starting material for calibrators and controls in LC/MS or GC/MS paroxetine testing applications in clinical toxicology, urine drug testing or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Paxil is a registered trademark of GlaxoSmithKline LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F

flash_point_c

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service