Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-012

Supelco

(±)-N-Desmethylselegiline solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N
CAS Number:
Molecular Weight:
173.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CC(NCC#C)CC1=CC=CC=C1

InChI

1S/C12H15N/c1-3-9-13-11(2)10-12-7-5-4-6-8-12/h1,4-8,11,13H,9-10H2,2H3

InChI key

UUFAJPMQSFXDFR-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. N-Desmethylselegiline, or norselegiline, is a major urinary metabolite of the antidepressant selegiline. Selegiline, which belongs to the phenethylamine class of drugs, is sold under the trade names Anipryl®, Emsam®, and Zelapar® for treatment of Parkinson′s disease, depression, and senile dementia.

Legal Information

Anipryl is a registered trademark of Deprenyl Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Emsam is a registered trademark of Somerset Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zelapar is a registered trademark of Valeant Pharmaceuticals North America LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debra S Harris et al.
BMC clinical pharmacology, 9, 13-13 (2009-08-04)
The selective MAO-B inhibitor selegiline has been evaluated in clinical trials as a potential medication for the treatment of cocaine dependence. This study evaluated the safety of and pharmacologic interactions between 7 days of transdermal selegiline dosed with patches (Selegiline
Kenichi Nishida et al.
Journal of analytical toxicology, 30(4), 232-237 (2006-06-29)
We devised a highly sensitive method for simultaneously determining methamphetamine (MA) and amphetamine (AP) enantiomers, desmethylselegiline (DMSG) and selegiline (SG), in human hair using a derivatization technique and high-performance liquid chromatography-electrospray ionization-mass spectrometry (HPLC-ESI-MS). MA and AP enantiomers and DMSG
Kazue Takahata et al.
European journal of pharmacology, 458(1-2), 81-89 (2002-12-25)
Selegiline, a therapeutic agent of Parkinson's disease, and its metabolite, desmethylselegiline, were explored for their neuroprotective effects against N-methyl-D-aspartate (NMDA)-induced cell death in rat retina. Morphometric analysis of the retina revealed that an intravitreal injection of NMDA induced a significant
K Magyar et al.
Journal of neural transmission. Supplementum, (72)(72), 165-173 (2007-11-07)
(-)-Deprenyl is a selective irreversible inhibitor of MAO-B. The parent compound is responsible for the enzyme inhibitory effect, but its metabolites are also playing a role in the complex pharmacological activity of the substance. In the present studies male NMRI
W A Baumgartner
Medical hypotheses, 54(5), 814-824 (2000-06-22)
The paper provides an interdisciplinary evaluation of the etiology, pathogenesis, and experimental treatments of retinitis pigmentosa (RP). It addresses a 10-year controversy concerning the rate of progression of RP. One laboratory has estimated remaining visual field to be lost at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service