Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDNLM-6786-10

Supelco

Aminomethylphosphonic acid-13C,15N,D2 solution

100 μg/mL in water, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CD2H415NO3P
CAS Number:
Molecular Weight:
115.04
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 10 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in water

format

single component solution

storage temp.

2-8°C

SMILES string

NCP(O)(O)=O

InChI

1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)

InChI key

MGRVRXRGTBOSHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service