Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-148

Supelco

Cholic acid-D4

100 μg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36D4O5
CAS Number:
Molecular Weight:
412.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H](CCC(O)=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C([2H])([2H])[C@H](O)C([2H])([2H])C[C@]4(C)[C@@]3([H])C[C@H](O)[C@@]21C

InChI

1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1/i8D2,10D2

InChI key

BHQCQFFYRZLCQQ-YFEOEUIKSA-N

General description

Cholic acid is a primary bile acid that plays an important role in cholesterol homeostasis and intestinal absorption of dietary vitamins and lipids. Levels of cholic acid as well as other bile acids serve as biomarkers for the diagnosis of numerous liver diseases from inborn errors of metabolism to cholangitis, cholestasis, and hepatitis. This Certified Spiking Solution® is suitable as an internal standard for quantitation of cholic acid levels in serum or other biological matrices by LC/MS or GC/MS for pharmaceutical research and clinical diagnostic testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service