Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-147

Supelco

Chenodeoxycholic acid-D4

100 μg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36D4O4
CAS Number:
Molecular Weight:
396.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC([2H])([2H])[C@@H](O)C3([2H])[2H])(C)[C@]3([H])C[C@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(O)=O

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1/i8D2,12D2

InChI key

RUDATBOHQWOJDD-PSTGXAJBSA-N

General description

Chenodeoxycholic acid (CDCA) is a primary bile acid that facilitates the intestinal uptake of lipids and fat-soluble vitamins. Levels of chenodeoxycholic acid can serve as markers for inborn errors of bile acid synthesis and hepatobiliary disorders such as familial intrahepatic cholestasis and biliary atresia. This stable-labeled internal standard is suitable for quantitation of CDCA levels in LC/MS and GC/MS bile acid testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service