Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-125

Supelco

Clozapine N-oxide solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4O
CAS Number:
Molecular Weight:
342.82
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C18H19ClN4O/c1-21-8-10-22(11-9-21)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)23(18)24/h2-7,12,24H,8-11H2,1H3

InChI key

YWSTWDVPYHSYLN-UHFFFAOYSA-N

General description

Clozapine N-oxide is the main metabolite of clozapine, an atypical antipsychotic prescribed for the treatment of schizophrenia and as an off-label treatment for bipolar disorder. This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service