Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-119

Supelco

Cocaine N-oxide hydrochloride

1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Cocaine N-oxide hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO5·HCl
Molecular Weight:
355.81
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in acetonitrile (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−70°C

SMILES string

Cl.COC(=O)C1C(C[C@@H]2CC[C@H]1[N+]2(C)[O-])OC(=O)c3ccccc3

InChI

1S/C17H21NO5.ClH/c1-18(21)12-8-9-13(18)15(17(20)22-2)14(10-12)23-16(19)11-6-4-3-5-7-11;/h3-7,12-15H,8-10H2,1-2H3;1H/t12-,13+,14?,15?,18?;/m0./s1

InChI key

JJKARSBCPADUTF-LMSTXNDJSA-N

General description

Cocaine N-oxide is a recently identified metabolite of cocaine that readily degrades to cocaine and norcocaine on GC/MS due to the compound′s sensitivity to heat. Testing laboratories are developing LC/MS methods for screening and quantitation of this metabolite to monitor cocaine use in matrices such as hair. Since this cocaine metabolite is listed as a controlled substance by US DEA, we provide this reference standard as a DEA-exempt solution for added convenience.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service